Order Entry
Northern Ireland
ContactUsLinkComponent
4-Fluoro-2-methylphenol
4-Fluoro-2-methylphenol
Catalog # APOSPC3823L-100G
Supplier:  Apollo Scientific
CAS Number:  
4-Fluoro-2-methylphenol
Catalog # APOSPC3823L-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:87 °C/14mmHg
Melting Pt:37…38 °C
Flash Pt:85 °C
 :126.13 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:452-72-2
Formula:C₇H₇FO
MDL Number:MFCD00075088