Order Entry
Northern Ireland
ContactUsLinkComponent
[(4-Fluorophenyl)sulphonyl]acetonitrile 97%
[(4-Fluorophenyl)sulphonyl]acetonitrile 97%
Catalog # APOSPC3487M-1G
Supplier:  Apollo Scientific
CAS Number:  
[(4-Fluorophenyl)sulphonyl]acetonitrile 97%
Catalog # APOSPC3487M-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :199.2 g/mol
Melting Pt:87…93 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:32083-66-2
Formula:C₈H₆FNO₂S
MDL Number:MFCD00179339