


3,5-Bis(trifluoromethyl)phenyl isothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23165-29-9
:
Formula:C₉H₃F₆NS
MDL Number:MFCD00040838
:271.19 g/mol
Boiling Pt:109…110 °C/35mmHg
Density:1,485 g/cm³ (25 °C)
Flash Pt:108 °C
UN:2922
ADR:8+6.1,III
:GHS05
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together