4,4'-Dimethyloctafluorobiphenyl 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26475-18-3
:
Formula:C₁₄H₆F₈
MDL Number:MFCD00000314
:326.19 g/mol
Melting Pt:148…149 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together