
3,4-Difluorobenzoyl chloride 98%
Catalog # APOSPC2727-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:176.55 g/mol
Boiling Pt:185…186 °C
Density:1,402 g/cm³ (25 °C)
Flash Pt:78 °C
Storage Temperature:Ambient
EINECS:278-570-7
UN:3265
ADR:8,II
:GHS05
CAS Number:76903-88-3
Formula:C₇H₃ClF₂O
MDL Number:MFCD00009930