Order Entry
Northern Ireland
ContactUsLinkComponent
4-Bromo-2-fluorobenzonitrile 98%
Irritant
4-Bromo-2-fluorobenzonitrile 98%
Catalog # APOSPC2665-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-2-fluorobenzonitrile 98%
Catalog # APOSPC2665-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :200.01 g/mol
Boiling Pt:238…239 °C
Melting Pt:69…72 °C
ADR:6.1,III
 :GHS07
Storage Temperature:Ambient
CAS Number:105942-08-3
Formula:C₇H₃BrFN
MDL Number:MFCD00143264