
1,2-Difluorobenzene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:367-11-3
:
Formula:C₆H₄F₂
MDL Number:MFCD00000284
:114.09 g/mol
Boiling Pt:92 °C
Melting Pt:–34 °C
Density:1,158 g/cm³ (25 °C)
Flash Pt:1 °C
Storage Temperature:Ambient
EINECS:206-680-7
UN:1993
ADR:3,II
:GHS02
Frequently Bought Together