Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole
APOSPC210005-1G
:
Apollo Scientific
:
2-Chloro-6-(trifluoromethyl)-1,3-benzoxazole
APOSPC210005-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
221.57 g/mol
:
GHS07
:
Refrigerator
:
1248393-79-4
:
C₈H₃ClF₃NO
:
MFCD18250485