

1-Bromo-2,6-difluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64248-56-2
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00009894
:192.99 g/mol
Boiling Pt:61 °C/35mmHg
Melting Pt:20 °C
Density:1,71 g/cm³ (25 °C)
Flash Pt:53 °C
Storage Temperature:Ambient
EINECS:264-750-2
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together