
Pentafluorophenyl-2-propanesulphonate
Catalog # APOSPC10303-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:886361-27-9
Formula:C₉H₇F₅O₃S
MDL Number:MFCD05975149