Order Entry
Northern Ireland
ContactUsLinkComponent
Triethylammonium dihydrogentrifluoride
Triethylammonium dihydrogentrifluoride
Catalog # APOSPC0945-500G
Supplier:  Apollo Scientific
CAS Number:  
Triethylammonium dihydrogentrifluoride
Catalog # APOSPC0945-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Nalgene bottle
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride
 :
 :161.21 g/mol
Melting Pt:–29 °C
EINECS:277-550-5
UN:2927
 :GHS05
 :GHS06
Storage Temperature:Refrigerator
CAS Number:73602-61-6
Formula:C₆H₁₈F₃N
MDL Number:MFCD00043294
Boiling Pt:70 °C/15mmHg
Density:0.989 g/cm³ (25 °C)
Flash Pt:87 °C
ADR:6.1+8 I