
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde
Catalog # APOSPC0941-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:170.139 g/mol
Melting Pt:114…118 °C
:GHS07
Storage Temperature:Ambient
CAS Number:79418-78-3
Formula:C₈H₇FO₃
MDL Number:MFCD02683560