

4-((Trifluoromethyl)thio)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:330-14-3
:
Formula:C₈H₄ClF₃OS
MDL Number:MFCD01631632
:240.63 g/mol
Boiling Pt:229…230 °C
Density:1,445 g/cm³ (25 °C)
Flash Pt:47,2 °C
Storage Temperature:Ambient
UN:2920
ADR:8+3,II
:GHS02
:GHS05
Frequently Bought Together