£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone
3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone 95%
3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone 95%
APOSOR950444-25G
:
Apollo Scientific
:
3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone 95%
APOSOR950444-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
178.19 g/mol
:
233 °C (dec.)
:
Ambient
:
3439
:
6.1,III
:
GHS06
:
28141-13-1
:
C₉H₁₀N₂O₂
:
MFCD00142624