Order Entry
Northern Ireland
ContactUsLinkComponent
2,4-Dinitro-1-naphthol 98%
Irritant
2,4-Dinitro-1-naphthol 98%
Catalog # APOSOR929998-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dinitro-1-naphthol 98%
Catalog # APOSOR929998-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :234.17 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:605-69-6
Formula:C₁₀H₆N₂O₅
MDL Number:MFCD00065352