Order Entry
Northern Ireland
ContactUsLinkComponent
Prop-2-ene-1-sulphonyl chloride 99%
Corrosive
Prop-2-ene-1-sulphonyl chloride 99%
Catalog # APOSOR918555-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Prop-2-ene-1-sulphonyl chloride 99%
Catalog # APOSOR918555-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :140.59 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
 :GHS05
CAS Number:14418-84-9
Formula:C₃H₅ClO₂S
MDL Number:MFCD08059789