Order Entry
Northern Ireland
ContactUsLinkComponent
1,2-Diphenylhydrazine 95%
1,2-Diphenylhydrazine 95%
Catalog # APOSOR904547-100G
Supplier:  Apollo Scientific
CAS Number:  
1,2-Diphenylhydrazine 95%
Catalog # APOSOR904547-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:122-66-7
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00003012