Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methylpiperidine
2-Methylpiperidine 99%
2-Methylpiperidine 99%
APOSOR8797-100ML
:
Apollo Scientific
:
2-Methylpiperidine 99%
APOSOR8797-100ML
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mL
Seq:
2440
:
:
99.18 g/mol
:
117…119 °C
:
–4 °C
:
0,844 g/cm³ (25 °C)
:
10 °C
:
2733
:
3+8,II
:
GHS02
:
GHS05
:
GHS07
:
Ambient
:
109-05-7
:
C₆H₁₃N
:
MFCD00005982