3-Methoxybenzylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5071-96-5
:
Formula:C₈H₁₁NO
MDL Number:MFCD00008115
:137.18 g/mol
Boiling Pt:230…231 °C
Density:1,7 g/cm³ (25 °C)
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:225-779-6
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together