Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Bromoisoquinoline
8-Bromoisoquinoline 95%
8-Bromoisoquinoline 95%
APOSOR8569-1G
:
Apollo Scientific
:
8-Bromoisoquinoline 95%
APOSOR8569-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
73…78 °C
:
208.0600 g/mol
:
GHS07
:
Ambient
:
63927-22-0
:
C₉H₆BrN
:
MFCD04973298