
11-Azido-3,6,9-trioxaundecan-1-amine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:134179-38-7
:
Formula:C₈H₁₈N₄O₃
MDL Number:MFCD00269874
:218.26 g/mol
Boiling Pt:198…202 °C
Density:1,1 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together