2'-Chloroacetanilide 98+%
Bottle, 5 g
Catalog # APOSOR7655-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: N-(2-Chlorophenyl)acetamide
- CAS Number:533-17-5
- Formula:C₈H₈ClNO
- MDL Number:MFCD00045169
- Molecular Weight:169.61 g/mol
- Melting Pt:87…89 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



