6-Amino-1,4-benzodioxane
:22013-33-8
:
:C₈H₉NO₂
:MFCD00006824
:151.17 g/mol
:116…118 °C/3mmHg
:26…31 °C
:1,231 g/cm³ (25 °C)
:120 °C
:244-718-4
:GHS07
:Refrigerator
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.