
Isopropylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15572-56-2
:
Formula:C₃H₉N
MDL Number:MFCD00050705
:95.57 g/mol
Melting Pt:139…150 °C
EINECS:239-629-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together