10-Bromodec-1-ene 95%
Catalog # APOSOR60224-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:219.1600 g/mol
Boiling Pt:50 °C/0,3mmHg
Density:1,092 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:62871-09-4
Formula:C₁₀H₁₉Br
MDL Number:MFCD00078660