9-Bromo-10-phenylanthracene
Catalog # APOSOR60163-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:333.23 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:23674-20-6
Formula:C₂₀H₁₃Br
MDL Number:MFCD00230983
Melting Pt:154…158 °C