Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-1-cycloheptene
1-Bromo-1-cycloheptene
1-Bromo-1-cycloheptene
APOSOR59406-1G
:
Apollo Scientific
:
1-Bromo-1-cycloheptene
APOSOR59406-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
81…82 °C/2mmHg
:
Refrigerator
:
18317-64-1
:
C₇H₁₁Br
:
MFCD09878419