D-(-)-Valine 98%
Catalog # APOSOR5884-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Melting Pt:>295 °C (subl.)
:117.15 g/mol
EINECS:211-368-9
:GHS07
Storage Temperature:Ambient
CAS Number:640-68-6
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064219
Melting Pt:>295 °C (subl.)