Order Entry
Northern Ireland
ContactUsLinkComponent
Cyclohexanethiol 97%
Cyclohexanethiol 97%
Catalog # APOSOR5294-25G
Supplier:  Apollo Scientific
CAS Number:  
Cyclohexanethiol 97%
Catalog # APOSOR5294-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :116.23 g/mol
Boiling Pt:158…160 °C
Melting Pt:–30 °C
Density:0,95 g/cm³ (25 °C)
Flash Pt:41 °C
Storage Temperature:Ambient
EINECS:216-378-7
UN:3054
ADR:3,III
 :GHS02
 :GHS07
CAS Number:1569-69-3
Formula:C₆H₁₂S
MDL Number:MFCD00001455