


Phthalic anhydride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85-44-9
:
Formula:C₈H₄O₃
MDL Number:MFCD00005918
:148.12 g/mol
Boiling Pt:284 °C
Melting Pt:131…134 °C
Density:1,53 g/cm³ (25 °C)
Flash Pt:152 °C
EINECS:201-607-5
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together