
Pyridine-2,6-dicarboxylic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:499-83-2
:
Formula:C₇H₅NO₄
MDL Number:MFCD00006299
:167.12 g/mol
Melting Pt:238…239 °C
Flash Pt:188 °C
EINECS:207-894-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together