Order Entry
Northern Ireland
ContactUsLinkComponent
1-Methyl-1H-1,2,4-triazole 97%
1-Methyl-1H-1,2,4-triazole 97%
Catalog # APOSOR480703-100G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-1H-1,2,4-triazole 97%
Catalog # APOSOR480703-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :83.10 g/mol
Boiling Pt:177…178 °C
Melting Pt:16 °C
Density:1,1 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:6086-21-1
Formula:C₃H₅N₃
MDL Number:MFCD01076192