L-Arginine methyl ester dihydrochloride 98%
Catalog # APOSOR471616-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:261.15 g/mol
Storage Temperature:Ambient
CAS Number:26340-89-6
Formula:C₇H₁₆N₄O₂
MDL Number:MFCD00038948