
2-Methyl-1-tetralone 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1590-08-5
:
Formula:C₁₁H₁₂O
MDL Number:MFCD00001690
:160.22 g/mol
Boiling Pt:127…131 °C/12mmHg
Density:1,057 g/cm³ (25 °C)
Flash Pt:>113 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together