Order Entry
Northern Ireland
ContactUsLinkComponent
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 98%
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 98%
Catalog # APOSOR46134-100G
Supplier:  Apollo Scientific
CAS Number:  
cis-5-Norbornene exo-2,3-dicarboxylic anhydride 98%
Catalog # APOSOR46134-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :164.16 g/mol
Melting Pt:140…145 °C
Storage Temperature:Ambient
CAS Number:2746-19-2
Formula:C₉H₈O₃
MDL Number:MFCD00630580