Order Entry
Northern Ireland
ContactUsLinkComponent
5-Amino-2-bromobenzoic acid
Toxic
5-Amino-2-bromobenzoic acid
Catalog # APOSOR4307-1G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
5-Amino-2-bromobenzoic acid
Catalog # APOSOR4307-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.03 g/mol
Melting Pt:171…179 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:2840-02-0
Formula:C₇H₆BrNO₂
MDL Number:MFCD07368968