

2-(Bromomethyl)naphthalene, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:939-26-4
:
Formula:C₁₁H₉Br
MDL Number:MFCD00004123
:221.1 g/mol
Boiling Pt:213 °C/100mmHg
Melting Pt:52…54 °C
Flash Pt:>110 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together