Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium methanesulphinate
Sodium methanesulphinate, Technical Grade
Sodium methanesulphinate, Technical Grade
APOSOR42101-25G
:
Apollo Scientific
:
Sodium methanesulphinate, Technical Grade
APOSOR42101-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
102.09 g/mol
:
222…226 °C (dec.)
:
243-669-6
:
GHS07
:
Ambient
:
20277-69-4
:
CH₃NaO₂S
:
MFCD00040392