

2,2,2-Trichloroethanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:115-20-8
:
Formula:C₂H₃Cl₃O
MDL Number:MFCD00004677
:149.4 g/mol
Boiling Pt:152…154 °C
Melting Pt:17,8 °C
Density:1,56 g/cm³ (25 °C)
Flash Pt:110 °C
Storage Temperature:Ambient
EINECS:204-071-0
:GHS07
:GHS05
Frequently Bought Together