Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
APOSOR350032-5G
:
Apollo Scientific
:
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate
APOSOR350032-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
185.22 g/mol
:
Ambient
:
35418-16-7
:
C₉H₁₅NO₃
:
MFCD06659481