4-(Methylsulphonyl)-2-nitrobenzoic acid
Catalog # APOSOR3369-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:245.2100 g/mol
Melting Pt:211…212 °C
:GHS07
Storage Temperature:Ambient
CAS Number:110964-79-9
Formula:C₈H₇NO₆S
MDL Number:MFCD01941299