Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Ureidobenzeneboronic acid pinacol ester
4-Ureidobenzeneboronic acid pinacol ester 98%
4-Ureidobenzeneboronic acid pinacol ester 98%
APOSOR3259-1G
:
Apollo Scientific
:
4-Ureidobenzeneboronic acid pinacol ester 98%
APOSOR3259-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
262.12 g/mol
:
124…128 °C
:
GHS07
:
Ambient
:
877134-77-5
:
C₁₃H₁₉BN₂O₃
:
MFCD06795683