1,3-Dimethyl-5-phenoxy-1H-pyrazole-4-carbaldehyde 95%
Catalog # APOSOR32557-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.24 g/mol
Storage Temperature:Ambient
CAS Number:109925-10-2
Formula:C₁₂H₁₂N₂O₂
MDL Number:MFCD07698583