
2,2-Dimethyl-1,3-dioxan-5-one
Catalog # APOSOR322571-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:130.14 g/mol
Boiling Pt:67 °C
Density:1,09 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
CAS Number:74181-34-3
Formula:C₆H₁₀O₃
MDL Number:MFCD00671514