Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-((Dimethylamino)methyl)aniline
4-((Dimethylamino)methyl)aniline
4-((Dimethylamino)methyl)aniline
4-((Dimethylamino)methyl)aniline
APOSOR322378-5G
:
Apollo Scientific
:
4-((Dimethylamino)methyl)aniline
APOSOR322378-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
150.2200 g/mol
:
Ambient
:
6406-74-2
:
C₉H₁₄N₂
:
MFCD00035953