1,8-Dibromonaphthalene 98%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:17135-74-9
- Formula:C₁₀H₆Br₂
- MDL Number:MFCD00183574
- Molecular Weight:285.97 g/mol
- Melting Pt:105…110 °C
- Storage Temperature:Refrigerator
- UN:3077
- ADR:9+EHS,III
3 Options Available Below
Product Details & Documents
- CAS Number:17135-74-9
- Formula:C₁₀H₆Br₂
- MDL Number:MFCD00183574
- Molecular Weight:285.97 g/mol
- Melting Pt:105…110 °C
- Storage Temperature:Refrigerator
- UN:3077
- ADR:9+EHS,III



