Order Entry
Northern Ireland
ContactUsLinkComponent
11-Bromo-1-undecanol
11-Bromo-1-undecanol
Catalog # APOSOR315399-100G
Supplier:  Apollo Scientific
CAS Number:  
11-Bromo-1-undecanol
Catalog # APOSOR315399-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :251.21 g/mol
Melting Pt:46…49 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1611-56-9
Formula:HO(CH₂)₁₀CH₂Br
MDL Number:MFCD00004752