
5-Bromo-6-methoxy-1H-indazole
Catalog # APOSOR30839-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.07 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:152626-78-3
Formula:C₈H₇BrN₂O
MDL Number:MFCD11007880