Order Entry
Northern Ireland
ContactUsLinkComponent
4-((tert-Butoxycarbonyl)amino)benzoic acid 98%
4-((tert-Butoxycarbonyl)amino)benzoic acid 98%
Catalog # APOSOR307084-100G
Supplier:  Apollo Scientific
CAS Number:  
4-((tert-Butoxycarbonyl)amino)benzoic acid 98%
Catalog # APOSOR307084-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :237.3 g/mol
Storage Temperature:Ambient
CAS Number:66493-39-8
Formula:C₁₂H₁₅NO₄
MDL Number:MFCD00037428