Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Indazole 98%

Indazole 98%

Irritant
Supplier:   Apollo Scientific
Indazole 98%
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  1H-Indazole
  • CAS Number:271-44-3
  • Formula:C₇H₆N₂
  • MDL Number:MFCD00005691
  • Molecular Weight:118.14 g/mol
  • Boiling Pt:270 °C/743mmHg
  • Melting Pt:145…149 °C
  • Storage Temperature:Ambient
  • EINECS:205-978-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £19.70
    £19.70
    1 * 5 g
       £19.70

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £56.40
    £56.40
    1 * 25 g
       £56.40

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.